Squaric Acid Dibutyl Ester, 97%, Thermo Scientific Chemicals
Squaric Acid Dibutyl Ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Squaric Acid Dibutyl Ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 133170050
Price (USD)/ Each
76.65
Online exclusive
90.20 
Save 13.55 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
76.65
Online exclusive
90.20 
Save 13.55 (15%)
Add to cart
Squaric Acid Dibutyl Ester, 97%, Thermo Scientific Chemicals
Catalog number133170050
Price (USD)/ Each
76.65
Online exclusive
90.20 
Save 13.55 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedibutoxycyclobut-3-ene-1,2-dione
Molecular FormulaC12H18O4
InChI KeyXBRWELTXMQSEIN-UHFFFAOYSA-N
SMILESCCCCOC1=C(OCCCC)C(=O)C1=O
Molecular Weight (g/mol)226.27
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Refractive index1.4930 to 1.4950 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only