Thermo Scientific Chemicals

Salicylsalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 133400250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Salicylsalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 133400250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 133400250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
552-94-3
IUPAC Name
2-(2-hydroxybenzoyloxy)benzoic acid
Molecular Formula
C14H10O5
InChI Key
WVYADZUPLLSGPU-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1OC(=O)C1=CC=CC=C1O
Titration with NaOH
>=97.5 %
Appearance (Color)
White
Infrared spectrum
Conforms
HPLC
>=97.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text