Salicylsalicylic acid, 98%, Thermo Scientific Chemicals
Salicylsalicylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Salicylsalicylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 133401000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-(2-hydroxybenzoyloxy)benzoic acid
Molecular FormulaC14H10O5
InChI KeyWVYADZUPLLSGPU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1OC(=O)C1=CC=CC=C1O
Molecular Weight (g/mol)258.23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH97.5 to 102.5 %
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only