1,1,2,2-Tetrachloroethane-d2 99 atom% D, Thermo Scientific Chemicals
1,1,2,2-Tetrachloroethane-d2 99 atom% D, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1,2,2-Tetrachloroethane-d2 99 atom% D, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
20 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 133580050
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.10 
Save 13.45 (15%)
-
Add to cart
Quantity:
5 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.10 
Save 13.45 (15%)
Add to cart
1,1,2,2-Tetrachloroethane-d2 99 atom% D, Thermo Scientific Chemicals
Catalog number133580050
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.10 
Save 13.45 (15%)
-
Add to cart
Chemical Identifiers
CAS33685-54-0
IUPAC Nametetrachloro(²H₂)ethane
Molecular FormulaC2H2Cl4
InChI KeyQPFMBZIOSGYJDE-QDNHWIQGSA-N
SMILES[2H]C(Cl)(Cl)C([2H])(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Water=<0.03 %
Appearance (Form)Liquid
D-Enrichment>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only