Nitric acid, extra pure, 60% solution in water, Thermo Scientific Chemicals
Nitric acid, extra pure, 60% solution in water, Thermo Scientific Chemicals
Nitric acid, extra pure, 60% solution in water, Thermo Scientific Chemicals
Nitric acid, extra pure, 60% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitric acid, extra pure, 60% solution in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Plastic bottle
Catalog number 133620025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration with NaOH59.0 to 63.0 %
Appearance (Color)Clear colorless to light yellow
Chloride (Cl)=<10 ppm
Iron (Fe)=<5 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link