Piperazine-2-carboxylic acid dihydrochloride, 98%, Thermo Scientific Chemicals
Piperazine-2-carboxylic acid dihydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Piperazine-2-carboxylic acid dihydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 133660100
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS3022-15-9
IUPAC Name(3R)-piperazine-1,4-diium-3-carboxylate
Molecular FormulaC5H11N2O2
InChI KeyJSSXHAMIXJGYCS-SCSAIBSYSA-O
SMILES[O-]C(=O)[C@H]1C[NH2+]CC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Titration Argentometric97.5 to 102.5 %
Titration with HClO497.5 to 102.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only