Ammonia, ca. 7N solution in methanol
Ammonia, ca. 7N solution in methanol
Ammonia, ca. 7N solution in methanol
Ammonia, ca. 7N solution in methanol
Thermo Scientific Chemicals

Ammonia, ca. 7N solution in methanol

Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger | CAS: 7664-41-7 | H3N | 17.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 133710010
Price (EUR)
168,30
Online Exclusive
203,00
Save 34,70 (17%)
Each
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
168,30
Online Exclusive
203,00
Save 34,70 (17%)
Each
Add to cart
Chemical Identifiers
CAS7664-41-7
IUPAC Nameammonia
Molecular FormulaH3N
InChI KeyQGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILESN
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 %
Strength6.0 to 7.5 N
Appearance (Form)Solution
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link