Thermo Scientific Chemicals

Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals

Catalog number: 133710251
25 L, Plastic Can, Each
Thermo Scientific Chemicals

Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals

Catalog number: 133710251
25 L, Plastic Can, Each
Quantity
Packaging
Catalog number: 133710251
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7664-41-7, 67-56-1
Appearance (Form)
Solution
Water
=<0.5 %
Strength
6.0 to 7.5 N
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text