(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals
(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog number 133730250
Price (USD)/ Each
-
Request bulk or custom format
Chemical Identifiers
CAS533-87-9
IUPAC Name9,10,16-trihydroxyhexadecanoic acid
Molecular FormulaC16H32O5
InChI KeyMEHUJCGAYMDLEL-UHFFFAOYNA-N
SMILESOCCCCCCC(O)C(O)CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Titration with NaOH>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only