4-Methylindole, 99%, Thermo Scientific Chemicals
4-Methylindole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylindole, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 133890010
Price (USD)/ Each
328.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
387.00 
Save 58.35 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
328.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
387.00 
Save 58.35 (15%)
Add to cart
4-Methylindole, 99%, Thermo Scientific Chemicals
Catalog number133890010
Price (USD)/ Each
328.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
387.00 
Save 58.35 (15%)
-
Add to cart
Chemical Identifiers
CAS16096-32-5
IUPAC Name4-methyl-1H-indole
Molecular FormulaC9H9N
InChI KeyPZOUSPYUWWUPPK-UHFFFAOYSA-N
SMILESCC1=C2C=CNC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Appearance (Form)Liquid after melting
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.6060 to 1.6080 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only