Thermo Scientific Chemicals

4-Iodotoluene, 98%, Thermo Scientific Chemicals

Catalog number: 133940500
50 g, Glass Bottle, Each
Thermo Scientific Chemicals

4-Iodotoluene, 98%, Thermo Scientific Chemicals

Catalog number: 133940500
50 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 133940500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
624-31-7
IUPAC Name
1-iodo-4-methylbenzene
Molecular Formula
C7H7I
InChI Key
UDHAWRUAECEBHC-UHFFFAOYSA-N
SMILES
CC1=CC=C(I)C=C1
Appearance (Form)
Low melting crystalline mass
Infrared spectrum
Conforms
Melting point
33°C to 36°C
GC
>=97.5 %
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text