Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
250 g
1 kg
2.5 kg
25 kg
Packaging:
View all
Plastic drum
Catalog number 134060100
Price (USD)/ Each
-
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Pellets
Titration with HCl>=85 %
Potassium carbonate=<1 % (K2CO3)
Chloride (Cl)=<70 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only