Thermo Scientific™
1300 Series Class II, Type A2 Biological Safety Cabinet Packages
Catalog number: 1342
Have Questions?
Thermo Scientific™
1300 Series Class II, Type A2 Biological Safety Cabinet Packages
Catalog number: 1342
Have Questions?
Perform daily applications safely and efficiently with simple-to-order Thermo Scientific™ 1300 Series Class II, Type A2 Biological Safety Cabinet. Package includes a cabinet with factory-installed UV light, manual adjustable height stand and one set of armrests. These safety cabinets deliver exceptional design and technology advancements such as superior protection with proprietary airflow design, exceptional ergonomics for a safe and comfortable environment, and outstanding energy efficiency for operational cost savings.
Catalog Number
1342
Unit Size
Each
Chamber Material
Stainless Steel (304)
Width (Metric) Exterior
100 cm
Sash Opening (Metric)
20.3 cm
Description
Width: 36 in.; 8 in. Window; Stainless-steel package; 120V 50/60Hz
Price (USD)
Full specifications
DescriptionWidth: 36 in.; 8 in. Window; Stainless-steel package; 120V 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level62 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20.3 cm
Width (English) Exterior39.4 in.
Volume (Metric) Exhaust15 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior100 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 39.4 x 61.8 in. (80 x 100 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior35.4 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
1323TS
also known as 1323
Unit Size
Each
Chamber Material
Stainless Steel (304)
Width (Metric) Exterior
100 cm
Sash Opening (Metric)
25.4 cm
Description
25.4 cm Window
Price (USD)
Full specifications
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level63 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior39.4 in.
Volume (Metric) Exhaust15 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior100 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 39.4 x 61.8 in. (80 x 100 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior35.4 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Chamber Material
Stainless Steel (304)
Width (Metric) Exterior
130 cm
Sash Opening (Metric)
20.3 cm
Description
20.3 cm Window
Price (USD)
Full specifications
Description20.3 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1300 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level62 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20.3 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust20 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior130 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 51.2 x 61.8 in. (80 x 130 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior51.2 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Chamber Material
Stainless Steel (304)
Width (Metric) Exterior
160 cm
Sash Opening (Metric)
20.3 cm
Description
20.3 cm Window
Price (USD)
Full specifications
Description20.3 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level64 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20.3 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Chamber Material
SmartCoat
Width (Metric) Exterior
160 cm
Sash Opening (Metric)
20.3 cm
Description
25.4 cm Window
Price (USD)
Full specifications
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20.3 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Chamber Material
Stainless Steel (304)
Width (Metric) Exterior
160 cm
Sash Opening (Metric)
25.4 cm
Description
25.4 cm Window
Price (USD)
Full specifications
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Chamber Material
Stainless Steel (304)
Width (Metric) Exterior
130 cm
Sash Opening (Metric)
25.4 cm
Description
25.4 cm Window
Price (USD)
Full specifications
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1300 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level63 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust20 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior130 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 51.2 x 61.8 in. (80 x 130 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior51.2 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Chamber Material
Stainless Steel (304)
Width (Metric) Exterior
190 cm
Sash Opening (Metric)
25.4 cm
Description
25.4 cm Window
Price (USD)
Full specifications
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust30 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior190 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 74.8 x 61.8 in. (80 x 190 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior74.8 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Chamber Material
SmartCoat
Width (Metric) Exterior
130 cm
Sash Opening (Metric)
25.4 cm
Description
25.4 cm Window
Price (USD)
Full specifications
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1300 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level63 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust20 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior130 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 51.2 x 61.8 in. (80 x 130 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior51.2 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Chamber Material
SmartCoat
Width (Metric) Exterior
190 cm
Sash Opening (Metric)
25.4 cm
Description
25.4 cm Window
Price (USD)
Full specifications
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust30 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior190 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 74.8 x 61.8 in. (80 x 190 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior74.8 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Showing 10 of 16Show more
Catalog NumberSpecificationsUnit SizeChamber MaterialWidth (Metric) ExteriorSash Opening (Metric)DescriptionPrice (USD)
1342Full specifications
EachStainless Steel (304)100 cm20.3 cmWidth: 36 in.; 8 in. Window; Stainless-steel package; 120V 50/60HzRequest A Quote
DescriptionWidth: 36 in.; 8 in. Window; Stainless-steel package; 120V 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level62 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20.3 cm
Width (English) Exterior39.4 in.
Volume (Metric) Exhaust15 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior100 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 39.4 x 61.8 in. (80 x 100 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior35.4 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1323TS
also known as 1323
Full specifications
EachStainless Steel (304)100 cm25.4 cm25.4 cm WindowRequest A Quote
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level63 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior39.4 in.
Volume (Metric) Exhaust15 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior100 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 39.4 x 61.8 in. (80 x 100 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior35.4 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1335Full specifications
EachStainless Steel (304)130 cm20.3 cm20.3 cm WindowRequest A Quote
Description20.3 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1300 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level62 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20.3 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust20 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior130 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 51.2 x 61.8 in. (80 x 130 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior51.2 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1353Full specifications
EachStainless Steel (304)160 cm20.3 cm20.3 cm WindowRequest A Quote
Description20.3 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level64 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20.3 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1369Full specifications
EachSmartCoat160 cm20.3 cm25.4 cm WindowRequest A Quote
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20.3 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1371Full specifications
EachStainless Steel (304)160 cm25.4 cm25.4 cm WindowRequest A Quote
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1375Full specifications
EachStainless Steel (304)130 cm25.4 cm25.4 cm WindowRequest A Quote
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1300 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level63 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust20 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior130 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 51.2 x 61.8 in. (80 x 130 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior51.2 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1377Full specifications
EachStainless Steel (304)190 cm25.4 cm25.4 cm WindowRequest A Quote
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust30 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior190 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 74.8 x 61.8 in. (80 x 190 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior74.8 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1385Full specifications
EachSmartCoat130 cm25.4 cm25.4 cm WindowRequest A Quote
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1300 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level63 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust20 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior130 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 51.2 x 61.8 in. (80 x 130 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior51.2 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1387Full specifications
EachSmartCoat190 cm25.4 cm25.4 cm WindowRequest A Quote
Description25.4 cm Window
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25.4 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust30 L
Filter Efficiency99.995% MPPS
Filter TypeH14 HEPA EN 1822, 99.995% MPPS
Width (Metric) Exterior190 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 74.8 x 61.8 in. (80 x 190 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior74.8 in.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Showing 10 of 16Show more
Benefits:
  • Save money over the entire life of the cabinet with energy efficient design.
  • Reduce operating costs up to 75% over traditional biological safety cabinets with AC motors.
  • Increase comfort and encourage safe working habits with the sloped front, bright workspace and low noise level.
  • Maximize safety through our innovative Thermo Scientific™ SmartFlow™ design with Digital Airflow Verification.
  • Organize the interior with confidence using SmartPort, a clean and easy way to organize with confidence due to the negative pressure dual-side wall.
  • Easily access the interior of the BSC by sliding down the front window.
Safety Features:
  • Series meets NSF/ANSI™ Standard 49.
  • Real-time airflow adjustments ensure inflow and downflow velocities remain steady.
  • Pressure sensor filter monitoring measures total airflow across the working area.
  • Proprietary SmartClean front window design simplifies cleaning for a safer working environment.
  • SmartFlow Indicator adjusts airflows as filter resistance changes.
  • SmartFlow visually demonstrates cabinet's ability to compensate to maintain personal protection.
More Choices:
  • Available in 3′, 4′, 5′ or 6′ widths (91.4 cm, 122 cm, 152.4 cm or 183 cm)
  • Available with either 8 in. or 10 in. work aperture (20.3 cm or 25.4 cm)
  • Available in SmartCoat or stainless-steel interior
  • Easy-to-order package configurations of popular accessories (manual adjustable height stand, UV light and armrest)
  • Electrical or manual adjustable height stands and Ergolign chair available
  • Build your own cabinet by selecting the cabinet-only part numbers and adding factory or field installed accessories.
Certifications/Compliance:
  • NSF/ANSI 49, UL, CE