5-Aminosalicylic acid, 99%, Thermo Scientific Chemicals
5-Aminosalicylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Aminosalicylic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 134330250
Price (USD)/ Each
27.10
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
27.10
Add to cart
5-Aminosalicylic acid, 99%, Thermo Scientific Chemicals
Catalog number134330250
Price (USD)/ Each
27.10
-
Add to cart
Chemical Identifiers
CAS89-57-6
IUPAC Name5-amino-2-hydroxybenzoic acid
Molecular FormulaC7H7NO3
InChI KeyKBOPZPXVLCULAV-UHFFFAOYSA-N
SMILESNC1=CC=C(O)C(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Color)Beige to grey to light brown
Titration with HClO4>=98.5 % (On dry substance)
Loss on drying=<0.5 % (1 g, 105°C, 3 hrs)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only