Activated charcoal, NORIT™ A SUPRA
Activated charcoal, NORIT™ A SUPRA
Activated charcoal, NORIT™ A SUPRA
Activated charcoal, NORIT™ A SUPRA
Thermo Scientific Chemicals

Activated charcoal, NORIT™ A SUPRA

Activated charcoal,NORIT™ A SUPRA, C, CAS Number-7440-44-0, acticarbone, activated charcoal, anthrasorb, carbon-12, carbono, carbosieve, graphene, graphite, mineral, norit, 1kg, Passes Test, CHEBI:27594, Black, 231-153-3, OKTJSMMVPCPJKN-UHFFFAOYSA-N, 15% max., 3652.0 deg.C, 15, 1807 | CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 134342500
Price (USD)
264.65
Special offer
Online exclusive
311.00
Save 46.35 (15%)
Each
In stock
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
264.65
Special offer
Online exclusive
311.00
Save 46.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Acid-soluble substances=<3 %
Adsorption power>=40 g/100 g (on Phenazone)
Methylene blue adsorption>=38 g/100 g
SulfidesPasses test
Alkali-soluble colored subst.Passes test
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection