Activated charcoal, NORIT(R) SA 2, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Activated charcoal, NORIT(R) SA 2, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
250 g
2.5 kg
Packaging:
View all
Fibre drum
Catalog number 134370250
Price (USD)/ Each
798.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
939.00 
Save 140.35 (15%)
-
Add to cart
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Price (USD)/ Each
798.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
939.00 
Save 140.35 (15%)
Add to cart
Activated charcoal, NORIT(R) SA 2, Thermo Scientific Chemicals
Catalog number134370250
Price (USD)/ Each
798.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
939.00 
Save 140.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Molasses number=<390
Appearance (Color)Black
Loss on drying=<10 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only