Thermo Scientific™
1300 Series A2 Class II, Type A2 Bio Safety Cabinets
Catalog number: 1347
Have Questions?
Thermo Scientific™
1300 Series A2 Class II, Type A2 Bio Safety Cabinets
Catalog number: 1347
Have Questions?
Perform daily applications safely and efficiently with Thermo Scientific™ 1300 Series A2 Class II, Type A2 Biosafety Cabinets. These biological safety cabinets deliver excellent design and technology advancements such as superior protection with proprietary airflow design, exceptional ergonomics for a safe and comfortable environment and outstanding energy efficiency for operational cost savings.
Catalog Number
1347
Unit Size
Each
Width (English) Cabinet
6 ft.
Chamber Material
Stainless Steel (304)
Sash Opening (English)
10 in.
Electrical Requirements
120 V, 50/60 Hz
Price (USD)
Full specifications
Description1347 - 6 ft wide, 10 inch window, Stainless Steel interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust30 L
Filter TypeHEPA
Width (Metric) Exterior190 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 74.8 x 61.8 in. (80 x 190 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior74.8 in.
Width (English) Cabinet6 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Width (English) Cabinet
4 ft.
Chamber Material
Stainless Steel (304)
Sash Opening (English)
8 in.
Electrical Requirements
120 V, 50/60 Hz
Price (USD)
Full specifications
Description1305 - 4 ft wide, 8 inch window, Stainless Steel interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1300 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level62 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust20 L
Filter TypeHEPA
Width (Metric) Exterior130 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 51.2 x 61.8 in. (80 x 130 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior51.2 in.
Width (English) Cabinet4 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Width (English) Cabinet
3 ft.
Chamber Material
Stainless Steel (304)
Sash Opening (English)
10 in.
Electrical Requirements
230 V, 50/60 Hz
Price (USD)
Full specifications
Description1332 - 3 ft wide, 10 inch window, Stainless Steel interior, Cabinet only - 230V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level63 dBA
Plug TypeCEE 7/7
Sash Opening (Metric)25 cm
Width (English) Exterior39.4 in.
Volume (Metric) Exhaust15 L
Filter TypeHEPA
Width (Metric) Exterior100 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 39.4 x 61.8 in. (80 x 100 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior35.4 in.
Width (English) Cabinet3 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Amperage7 A to 8 A
Unit SizeEach
Catalog Number
Unit Size
Each
Width (English) Cabinet
3 ft.
Chamber Material
Stainless Steel (304)
Sash Opening (English)
8 in.
Electrical Requirements
120 V, 50/60 Hz
Price (USD)
Full specifications
Description1341 - 3 ft wide, 8 inch window, Stainless Steel interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level62 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20 cm
Width (English) Exterior39.4 in.
Volume (Metric) Exhaust15 L
Filter TypeHEPA
Width (Metric) Exterior100 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 39.4 x 61.8 in. (80 x 100 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior35.4 in.
Width (English) Cabinet3 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Width (English) Cabinet
5 ft.
Chamber Material
Stainless Steel (304)
Sash Opening (English)
8 in.
Electrical Requirements
120 V, 50/60 Hz
Price (USD)
Full specifications
Description1352 - 5 ft wide, 8 inch window, Stainless Steel interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level64 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter TypeHEPA
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Width (English) Cabinet5 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
Unit Size
Each
Width (English) Cabinet
4 ft.
Chamber Material
SmartCoat
Sash Opening (English)
10 in.
Electrical Requirements
230 V, 50/60 Hz
Price (USD)
Full specifications
Description1356 - 4 ft wide, 10 inch window, SmartCoat interior, Cabinet only - 230V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1300 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level63 dBA
Plug TypeCEE 7/7
Sash Opening (Metric)25 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust20 L
Filter TypeHEPA
Width (Metric) Exterior130 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 51.2 x 61.8 in. (80 x 130 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior51.2 in.
Width (English) Cabinet4 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Amperage7 A to 8 A
Unit SizeEach
Catalog Number
Unit Size
Each
Width (English) Cabinet
6 ft.
Chamber Material
SmartCoat
Sash Opening (English)
10 in.
Electrical Requirements
120 V, 50/60 Hz
Price (USD)
Full specifications
Description1357 - 6 ft wide, 10 inch window, SmartCoat interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust30 L
Filter TypeHEPA
Width (Metric) Exterior190 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 74.8 x 61.8 in. (80 x 190 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior74.8 in.
Width (English) Cabinet6 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Catalog Number
1358TS
also known as 1358
Unit Size
Each
Width (English) Cabinet
6 ft.
Chamber Material
SmartCoat
Sash Opening (English)
10 in.
Electrical Requirements
230 V, 50/60 Hz
Price (USD)
Full specifications
Description1358 - 6 ft wide, 10 inch window, SmartCoat interior, Cabinet only - 230V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeCEE 7/7
Sash Opening (Metric)25 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust30 L
Filter TypeHEPA
Width (Metric) Exterior190 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 74.8 x 61.8 in. (80 x 190 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior74.8 in.
Width (English) Cabinet6 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Amperage7 A to 8 A
Unit SizeEach
Catalog Number
Unit Size
Each
Width (English) Cabinet
5 ft.
Chamber Material
Stainless Steel (304)
Sash Opening (English)
8 in.
Electrical Requirements
230 V, 50/60 Hz
Price (USD)
Full specifications
Description1366 - 5 ft wide, 8 inch window, Stainless Steel interior, Cabinet only - 230V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level64 dBA
Plug TypeCEE 7/7
Sash Opening (Metric)20 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter TypeHEPA
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Width (English) Cabinet5 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Amperage7 A to 8 A
Unit SizeEach
Catalog Number
Unit Size
Each
Width (English) Cabinet
5 ft.
Chamber Material
SmartCoat
Sash Opening (English)
10 in.
Electrical Requirements
120 V, 50/60 Hz
Price (USD)
Full specifications
Description1368 - 5 ft wide, 10 inch window, SmartCoat interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter TypeHEPA
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Width (English) Cabinet5 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Showing 10 of 28Show more
Catalog NumberSpecificationsUnit SizeWidth (English) CabinetChamber MaterialSash Opening (English)Electrical RequirementsPrice (USD)
1347Full specifications
Each6 ft.Stainless Steel (304)10 in.120 V, 50/60 HzRequest A Quote
Description1347 - 6 ft wide, 10 inch window, Stainless Steel interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust30 L
Filter TypeHEPA
Width (Metric) Exterior190 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 74.8 x 61.8 in. (80 x 190 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior74.8 in.
Width (English) Cabinet6 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1305Full specifications
Each4 ft.Stainless Steel (304)8 in.120 V, 50/60 HzRequest A Quote
Description1305 - 4 ft wide, 8 inch window, Stainless Steel interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1300 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level62 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust20 L
Filter TypeHEPA
Width (Metric) Exterior130 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 51.2 x 61.8 in. (80 x 130 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior51.2 in.
Width (English) Cabinet4 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1332Full specifications
Each3 ft.Stainless Steel (304)10 in.230 V, 50/60 HzRequest A Quote
Description1332 - 3 ft wide, 10 inch window, Stainless Steel interior, Cabinet only - 230V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level63 dBA
Plug TypeCEE 7/7
Sash Opening (Metric)25 cm
Width (English) Exterior39.4 in.
Volume (Metric) Exhaust15 L
Filter TypeHEPA
Width (Metric) Exterior100 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 39.4 x 61.8 in. (80 x 100 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior35.4 in.
Width (English) Cabinet3 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Amperage7 A to 8 A
Unit SizeEach
1341Full specifications
Each3 ft.Stainless Steel (304)8 in.120 V, 50/60 HzRequest A Quote
Description1341 - 3 ft wide, 8 inch window, Stainless Steel interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level62 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20 cm
Width (English) Exterior39.4 in.
Volume (Metric) Exhaust15 L
Filter TypeHEPA
Width (Metric) Exterior100 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 39.4 x 61.8 in. (80 x 100 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior35.4 in.
Width (English) Cabinet3 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1352Full specifications
Each5 ft.Stainless Steel (304)8 in.120 V, 50/60 HzRequest A Quote
Description1352 - 5 ft wide, 8 inch window, Stainless Steel interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level64 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)20 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter TypeHEPA
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Width (English) Cabinet5 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1356Full specifications
Each4 ft.SmartCoat10 in.230 V, 50/60 HzRequest A Quote
Description1356 - 4 ft wide, 10 inch window, SmartCoat interior, Cabinet only - 230V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1300 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level63 dBA
Plug TypeCEE 7/7
Sash Opening (Metric)25 cm
Width (English) Exterior51.2 in.
Volume (Metric) Exhaust20 L
Filter TypeHEPA
Width (Metric) Exterior130 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 51.2 x 61.8 in. (80 x 130 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior51.2 in.
Width (English) Cabinet4 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Amperage7 A to 8 A
Unit SizeEach
1357Full specifications
Each6 ft.SmartCoat10 in.120 V, 50/60 HzRequest A Quote
Description1357 - 6 ft wide, 10 inch window, SmartCoat interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust30 L
Filter TypeHEPA
Width (Metric) Exterior190 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 74.8 x 61.8 in. (80 x 190 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior74.8 in.
Width (English) Cabinet6 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
1358TS
also known as 1358
Full specifications
Each6 ft.SmartCoat10 in.230 V, 50/60 HzRequest A Quote
Description1358 - 6 ft wide, 10 inch window, SmartCoat interior, Cabinet only - 230V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1900 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeCEE 7/7
Sash Opening (Metric)25 cm
Width (English) Exterior74.8 in.
Volume (Metric) Exhaust30 L
Filter TypeHEPA
Width (Metric) Exterior190 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 74.8 x 61.8 in. (80 x 190 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior74.8 in.
Width (English) Cabinet6 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Amperage7 A to 8 A
Unit SizeEach
1366Full specifications
Each5 ft.Stainless Steel (304)8 in.230 V, 50/60 HzRequest A Quote
Description1366 - 5 ft wide, 8 inch window, Stainless Steel interior, Cabinet only - 230V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level64 dBA
Plug TypeCEE 7/7
Sash Opening (Metric)20 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter TypeHEPA
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)8 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Width (English) Cabinet5 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Amperage7 A to 8 A
Unit SizeEach
1368Full specifications
Each5 ft.SmartCoat10 in.120 V, 50/60 HzRequest A Quote
Description1368 - 5 ft wide, 10 inch window, SmartCoat interior, Cabinet only - 115V, 50/60Hz
Certifications/ComplianceNSF/ANSI 49, UL, CE
Chamber MaterialSmartCoat
Width (Metric) Interior1500 mm
Product TypeClass II, Type A2 Bio Safety Cabinet
Model1300 Series
Noise Level65 dBA
Plug TypeNEMA 5-15
Sash Opening (Metric)25 cm
Width (English) Exterior63 in.
Volume (Metric) Exhaust25 L
Filter TypeHEPA
Width (Metric) Exterior160 cm
Ports1 x Each Side Panel
Load Bearing Capacity (English)110 lb.
Load Bearing Capacity (Metric)50 Kg
Sash Opening (English)10 in.
Service Fixtures6 (3 on Each Side)
Dimensions (L x W x H) Exterior31.5 x 63 x 61.8 in. (80 x 160 x 156.8 cm)
Length (English) Exterior31.5 in.
Length (Metric) Exterior80 cm
Width (English) Interior59.1 in.
Width (English) Cabinet5 ft.
Frequency50/60 Hz
Height (English) Exterior61.8 in.
Height (English) Max. Opening21 in.
Height (Metric) Exterior156.8 cm
Height (Metric) Max. Opening53.5 cm
Electrical Requirements120 V, 50/60 Hz
Voltage120 V
Amperage8 A to 10 A
Unit SizeEach
Showing 10 of 28Show more
Features
  • Save money over the entire life of the cabinet with energy-efficient design
  • Reduce operating costs up to 75% over traditional biological safety cabinets with AC motors
  • Increase comfort and encourage safe working habits with the sloped front, bright workspace and low noise level
  • Maximize safety through the innovative Thermo Scientific™ SmartFlow™ design with Digital Airflow Verification
Safety Features:
  • Cabinet meets NSF/ANSI™ Standard 49.
  • Real-time airflow adjustments ensure inflow and downflow velocities remain steady.
  • Pressure sensor filter monitoring measures total airflow across the working area.
  • Proprietary front window design simplifies cleaning for a safer working environment.
  • SmartFlow Indicator adjusts airflows as filter resistance changes and visually demonstrates cabinet's ability to compensate to maintain personal protection.
More Choices:
  • Available in 3, 4, 5, or 6' widths (0.9 m, 1.2 m, 1.5 m, or 1.8 m)
  • Available with either 8 or 10 in. work aperture (20.3 or 25.4cm)
  • Available in SmartCoat or stainless-steel interior
  • Electrical or manual adjustable height stands available (must be ordered separately)
Certifications/Compliance:
  • NSF/ANSI 49, UL, CE