4-Cyano-4-phenylcyclohexanone, 97%, Thermo Scientific Chemicals
4-Cyano-4-phenylcyclohexanone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyano-4-phenylcyclohexanone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number 134900050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS25115-74-6
IUPAC Name4-oxo-1-phenylcyclohexane-1-carbonitrile
Molecular FormulaC13H13NO
InChI KeyGKXOABVSZWCJJK-UHFFFAOYSA-N
SMILESO=C1CCC(CC1)(C#N)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline solid
Infrared spectrumConforms
Melting point114°C to 119°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only