2-Methyl-1-pyrroline, 98%, Thermo Scientific Chemicals
2-Methyl-1-pyrroline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-1-pyrroline, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 135070250
Price (USD)/ Each
134.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
158.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
134.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
158.00 
Save 23.35 (15%)
Add to cart
2-Methyl-1-pyrroline, 98%, Thermo Scientific Chemicals
Catalog number135070250
Price (USD)/ Each
134.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
158.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-methylpyrrolidin-2-ylium
Molecular FormulaC5H10N
InChI KeyOZLJZWIVBYLSOK-UHFFFAOYSA-N
SMILESC[C+]1CCCN1
Molecular Weight (g/mol)84.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4400 to 1.4420 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only