D(-)-Glutamine 98%
Thermo Scientific Chemicals

D(-)-Glutamine 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 135160050
Price (USD)/ Each
229.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
270.00 
Save 40.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
229.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
270.00 
Save 40.35 (15%)
Add to cart
D(-)-Glutamine 98%
Catalog number135160050
Price (USD)/ Each
229.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
270.00 
Save 40.35 (15%)
-
Add to cart
Chemical Identifiers
CAS5959-95-5
IUPAC Name(2R)-2-amino-4-carbamoylbutanoic acid
Molecular FormulaC5H10N2O3
InChI KeyZDXPYRJPNDTMRX-GSVOUGTGSA-N
SMILESN[C@H](CCC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Specific optical rotation-30° to -34° (20°C, 589 nm) (c=10, 2 N HCl)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only