Thermo Scientific Chemicals

Glycidyl phenyl ether, 99%, Thermo Scientific Chemicals

Catalog number: 135352500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Glycidyl phenyl ether, 99%, Thermo Scientific Chemicals

Catalog number: 135352500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 135352500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
122-60-1
IUPAC Name
2-(phenoxymethyl)oxirane
Molecular Formula
C9H10O2
InChI Key
FQYUMYWMJTYZTK-UHFFFAOYNA-N
SMILES
C(OC1=CC=CC=C1)C1CO1
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.5295 to 1.5315 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text