Isopropyltriphenylphosphonium iodide, 98%, Thermo Scientific Chemicals
Isopropyltriphenylphosphonium iodide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropyltriphenylphosphonium iodide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 137080250
Price (USD)/ Each
79.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
93.40 
Save 13.75 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
79.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
93.40 
Save 13.75 (15%)
Add to cart
Isopropyltriphenylphosphonium iodide, 98%, Thermo Scientific Chemicals
Catalog number137080250
Price (USD)/ Each
79.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
93.40 
Save 13.75 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametriphenyl(propan-2-yl)phosphanium iodide
Molecular FormulaC21H22IP
InChI KeyHHBXWXJLQYJJBW-UHFFFAOYSA-M
SMILES[I-].CC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)432.29
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point195°C to 200°C
Titration Argentometric97.5 to 102.5 %
Appearance (Form)Powder or crystalline powder or crystals
Appearance (Color)Light yellow to beige to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only