Ethyl 3-mercaptopropionate, 98%, Thermo Scientific Chemicals
Ethyl 3-mercaptopropionate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3-mercaptopropionate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 137410250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5466-06-8
IUPAC Nameethyl 3-sulfanylpropanoate
Molecular FormulaC5H10O2S
InChI KeyCJQWLNNCQIHKHP-UHFFFAOYSA-N
SMILESCCOC(=O)CCS
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4560 to 1.4580 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only