Thermo Scientific Chemicals

D(-)-Tartaric Acid (unnatural Tartaric Acid) 99%, Thermo Scientific Chemicals

Catalog number: 137861000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

D(-)-Tartaric Acid (unnatural Tartaric Acid) 99%, Thermo Scientific Chemicals

Catalog number: 137861000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 137861000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
147-71-7
IUPAC Name
2,3-dihydroxybutanedioic acid
Molecular Formula
C4H6O6
InChI Key
FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES
OC(C(O)C(O)=O)C(O)=O
Heavy metals (as Pb)
=<10 ppm
Sulfated ash
=<0.1 %
Specific optical rotation
-11.0° to -13.0° (20°C, 589 nm) (c=20, water)
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text