DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 137875000
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
Add to cart
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
Catalog number137875000
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS133-37-9
IUPAC Name2,3-dihydroxybutanedioic acid
Molecular FormulaC4H6O6
InChI KeyFEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILESOC(C(O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point200°C to 206°C
Titration with NaOH>=99.4 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only