Thermo Scientific Chemicals

p-Terphenyl, 99+%, pure

Catalog number: 137921000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

p-Terphenyl, 99+%, pure

Catalog number: 137921000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 137921000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92-94-4
IUPAC Name
4-phenyl-1,1'-biphenyl
Molecular Formula
C18H14
InChI Key
XJKSTNDFUHDPQJ-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Appearance (Color)
White to light beige
Appearance (Form)
flakes
Infrared spectrum
Conforms
Melting point
211°C to 216°C
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text