Thermo Scientific Chemicals

2,4,5,6-Tetraaminopyrimidine sulfate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 137930250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4,5,6-Tetraaminopyrimidine sulfate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 137930250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 137930250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5392-28-9
IUPAC Name
pyrimidine-2,4,5,6-tetramine; sulfuric acid
Molecular Formula
C4H10N6O4S
InChI Key
MQEFDQWUCTUJCP-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 % (On dry substance)
Loss on drying
=<9 % (1 g, 150°C, 3 hrs)
Appearance (Color)
Ochre to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text