Thermo Scientific Chemicals

alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%, Thermo Scientific Chemicals

Catalog number: 137971000
100 g, Each
Thermo Scientific Chemicals

alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%, Thermo Scientific Chemicals

Catalog number: 137971000
100 g, Each
Quantity
Catalog number: 137971000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13209-15-9
IUPAC Name
1,2-bis(dibromomethyl)benzene
Molecular Formula
C8H6Br4
InChI Key
LNAOKZKISWEZNY-UHFFFAOYSA-N
SMILES
BrC(Br)C1=CC=CC=C1C(Br)Br
Melting point
114°C to 118°C
GC
>=96.0 %
Appearance (Color)
Off-white to beige to light grey
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text