Tetra-n-butyltin, 96%, Thermo Scientific Chemicals
Tetra-n-butyltin, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetra-n-butyltin, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 137980050
Price (USD)/ Each
17.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
20.90 
Save 3.25 (16%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
17.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
20.90 
Save 3.25 (16%)
Add to cart
Tetra-n-butyltin, 96%, Thermo Scientific Chemicals
Catalog number137980050
Price (USD)/ Each
17.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
20.90 
Save 3.25 (16%)
-
Add to cart
Chemical Identifiers
CAS1461-25-2
IUPAC Nametetrabutylstannane
Molecular FormulaC16H36Sn
InChI KeyAFCAKJKUYFLYFK-UHFFFAOYSA-N
SMILESCCCC[Sn](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=95 %
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to straw yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only