Thermo Scientific Chemicals

Tetra-n-butyltin, 96%, Thermo Scientific Chemicals

Catalog number: 137985000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetra-n-butyltin, 96%, Thermo Scientific Chemicals

Catalog number: 137985000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 137985000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1461-25-2
IUPAC Name
tetrabutylstannane
Molecular Formula
C16H36Sn
InChI Key
AFCAKJKUYFLYFK-UHFFFAOYSA-N
SMILES
CCCC[Sn](CCCC)(CCCC)CCCC
GC
>=95 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to straw yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text