Tetrachloroethylene, 99%, extra pure, Thermo Scientific Chemicals
Tetrachloroethylene, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrachloroethylene, 99%, extra pure, Thermo Scientific Chemicals

Tetrachloroethylene, 99%, C2Cl4, CAS Number-127-18-4, ankilostin, carbon dichloride, ethene, tetrachloro, ethylene tetrachloride, perc, perchlorethylene, perchloroethylene, tetrachlorethylene, tetrachloroethene, tetrachloroethylene, 2.5L, 0.003% max. (NaOH), 98.5% min. (GC), 01, 187
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 138010025
Price (USD)/ Each
186.65
Online exclusive
207.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
186.65
Online exclusive
207.00 
Save 20.35 (10%)
Add to cart
Tetrachloroethylene, 99%, extra pure, Thermo Scientific Chemicals
Catalog number138010025
Price (USD)/ Each
186.65
Online exclusive
207.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS127-18-4
IUPAC Nametetrachloroethene
Molecular FormulaC2Cl4
InChI KeyCYTYCFOTNPOANT-UHFFFAOYSA-N
SMILESClC(Cl)=C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<15 APHA
Water=<0.01 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only