Thermo Scientific Chemicals

Tetracyanoethylene, 98%, Thermo Scientific Chemicals

Catalog number: 138050100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetracyanoethylene, 98%, Thermo Scientific Chemicals

Catalog number: 138050100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 138050100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
670-54-2
IUPAC Name
eth-1-ene-1,1,2,2-tetracarbonitrile
Molecular Formula
C6N4
InChI Key
NLDYACGHTUPAQU-UHFFFAOYSA-N
SMILES
N#CC(C#N)=C(C#N)C#N
Infrared spectrum
Conforms
Melting point
196°C to 201°C
Appearance (Form)
Crystalline powder or crystals and/or chunks
Assay
>=97.5 % (Ex N)
Appearance (Color)
White to beige to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text