Thermo Scientific Chemicals

1-Tetradecylamine, 98%, Thermo Scientific Chemicals

Catalog number: 138100500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Tetradecylamine, 98%, Thermo Scientific Chemicals

Catalog number: 138100500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 138100500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2016-42-4
IUPAC Name
hydrogen tetradecan-1-amine chloride
Molecular Formula
C14H32ClN
InChI Key
KKWUACQXLWHLCX-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CCCCCCCCCCCCCCN
Appearance (Color)
White to pink
Titration with HClO4
>=97.5 %
Appearance (Form)
Crystals or crystalline solid
Infrared spectrum
Conforms
Melting point
37°C to 42°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text