Tetraethylenepentamine, tech., Thermo Scientific Chemicals
Tetraethylenepentamine, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetraethylenepentamine, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 138112500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-57-2
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Titration with HCl>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only