1,2,3,4-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals
1,2,3,4-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2,3,4-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 138271000
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.60 
Save 8.95 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.60 
Save 8.95 (15%)
Add to cart
1,2,3,4-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals
Catalog number138271000
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.60 
Save 8.95 (15%)
-
Add to cart
Chemical Identifiers
CAS635-46-1
IUPAC Name1,2,3,4-tetrahydroquinoline
Molecular FormulaC9H11N
InChI KeyLBUJPTNKIBCYBY-UHFFFAOYSA-N
SMILESC1CNC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
AppearanceMay darken on storage
Refractive index1.5930 to 1.5960 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear yellow to orange
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only