α-Tetralone, 98%, Thermo Scientific Chemicals
α-Tetralone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

α-Tetralone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 138310250
Price (USD)/ Each
29.90
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
29.90
Add to cart
α-Tetralone, 98%, Thermo Scientific Chemicals
Catalog number138310250
Price (USD)/ Each
29.90
-
Add to cart
Chemical Identifiers
CAS529-34-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to amber to brown
Appearance (Form)Liquid
AppearanceMay darken on storage
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only