Thermo Scientific Chemicals

Tetramethylammonium chloride, 98+%, Thermo Scientific Chemicals

Catalog number: 138330010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Tetramethylammonium chloride, 98+%, Thermo Scientific Chemicals

Catalog number: 138330010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 138330010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-57-0
IUPAC Name
tetramethylazanium chloride
Molecular Formula
C4H12ClN
InChI Key
OKIZCWYLBDKLSU-UHFFFAOYSA-M
SMILES
[Cl-].C[N+](C)(C)C
Appearance (Color)
White to cream
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Titration Argentometric
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text