Thermo Scientific Chemicals

1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals

Catalog number: 138371000
100 g, Each
Thermo Scientific Chemicals

1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals

Catalog number: 138371000
100 g, Each
Quantity
Catalog number: 138371000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
95-93-2
IUPAC Name
1,2,4,5-tetramethylbenzene
Molecular Formula
C10H14
InChI Key
SQNZJJAZBFDUTD-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(C)C=C1C
GC
>=97.0 %
Appearance (Form)
Crystals
Melting point
78°C to 82°C
Appearance (Color)
Semitransparent white to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text