Thermo Scientific Chemicals

Tetramethylene sulfone, 99+%, Thermo Scientific Chemicals

Catalog number: 138430010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Tetramethylene sulfone, 99+%, Thermo Scientific Chemicals

Catalog number: 138430010
1 L, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 138430010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
126-33-0
IUPAC Name
1λ⁶-thiolane-1,1-dione
Molecular Formula
C4H8O2S
InChI Key
HXJUTPCZVOIRIF-UHFFFAOYSA-N
SMILES
O=S1(=O)CCCC1
Infrared spectrum
Conforms
Water
=<0.3 %
Appearance (Form)
Liquid after melting
Refractive index
1.4820 to 1.4850 (20°C, 589 nm)
Appearance (Color)
Clear yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text