Tetramethylene sulfone, 99+%, Thermo Scientific Chemicals
Tetramethylene sulfone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetramethylene sulfone, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 138430025
Price (USD)/ Each
279.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
329.00 
Save 49.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
279.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
329.00 
Save 49.35 (15%)
Add to cart
Tetramethylene sulfone, 99+%, Thermo Scientific Chemicals
Catalog number138430025
Price (USD)/ Each
279.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
329.00 
Save 49.35 (15%)
-
Add to cart
Chemical Identifiers
CAS126-33-0
IUPAC Name1λ⁶-thiolane-1,1-dione
Molecular FormulaC4H8O2S
InChI KeyHXJUTPCZVOIRIF-UHFFFAOYSA-N
SMILESO=S1(=O)CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.3 %
Appearance (Form)Liquid after melting
Refractive index1.4820 to 1.4850 (20°C, 589 nm)
Appearance (Color)Clear yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only