2,6,10,14-Tetramethylpentadecane, 95%, Thermo Scientific Chemicals
2,6,10,14-Tetramethylpentadecane, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6,10,14-Tetramethylpentadecane, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
50 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 138462500
Price (USD)/ Each
824.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
970.00 
Save 145.35 (15%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
824.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
970.00 
Save 145.35 (15%)
Add to cart
2,6,10,14-Tetramethylpentadecane, 95%, Thermo Scientific Chemicals
Catalog number138462500
Price (USD)/ Each
824.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
970.00 
Save 145.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,6,10,14-tetramethylpentadecane
Molecular FormulaC19H40
InChI KeyXOJVVFBFDXDTEG-UHFFFAOYNA-N
SMILESCC(C)CCCC(C)CCCC(C)CCCC(C)C
Molecular Weight (g/mol)268.53
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94.0 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only