Thermo Scientific Chemicals

Tetramethylsilane, 99.9+%, NMR grade, Thermo Scientific Chemicals

Catalog number: 138470250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetramethylsilane, 99.9+%, NMR grade, Thermo Scientific Chemicals

Catalog number: 138470250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 138470250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-76-3
IUPAC Name
tetramethylsilane
Molecular Formula
C4H12Si
InChI Key
CZDYPVPMEAXLPK-UHFFFAOYSA-N
SMILES
C[Si](C)(C)C
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
GC
>=99.90 %
Suitability
(for 1H-NMR position ref.) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text