1,1,3,3-Tetramethylurea, 99%, Thermo Scientific Chemicals
1,1,3,3-Tetramethylurea, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1,3,3-Tetramethylurea, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
250 mL
Packaging:
Glass bottle
Catalog number 138490010
Price (USD)/ Each
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS632-22-4
IUPAC Name1,1,3,3-tetramethylurea
Molecular FormulaC5H12N2O
InChI KeyAVQQQNCBBIEMEU-UHFFFAOYSA-N
SMILESCN(C)C(=O)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Water=<0.5 % (K.F.)
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only