Tetraphenylboron sodium, 99.5%, Thermo Scientific Chemicals
Tetraphenylboron sodium, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetraphenylboron sodium, 99.5%, Thermo Scientific Chemicals

Tetraphenylboron sodium, 99.5%, C24H20BNa, CAS Number-143-66-8, borate 1-, tetraphenyl-, sodium, dotite kalibor, kalibor, kalignost, kariporu k, sodium tetraphenylborate, sodium tetraphenylborate 1-, sodium tetraphenylboride, sodium tetraphenylboron, tetraphenylboron sodium, 100g
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 138520250
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
Add to cart
Tetraphenylboron sodium, 99.5%, Thermo Scientific Chemicals
Catalog number138520250
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS143-66-8
IUPAC Namesodium tetraphenylboranuide
Molecular FormulaC24H20BNa
InChI KeyHFSRCEJMTLMDLI-UHFFFAOYSA-N
SMILES[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Titration with HClO499.4 to 100.6 %
Infrared spectrumConforms
Loss on drying=<0.5 % (105°C)
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only