Thermo Scientific Chemicals

Thenoyltrifluoroacetone, 99%, Thermo Scientific Chemicals

Catalog number: 138615000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Thenoyltrifluoroacetone, 99%, Thermo Scientific Chemicals

Catalog number: 138615000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 138615000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
326-91-0
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Appearance (Color)
Light yellow to beige to brown
Melting point
40°C to 44°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text