Thermo Scientific Chemicals

Thianaphthene, 97%, Thermo Scientific Chemicals

Catalog number: 138620250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Thianaphthene, 97%, Thermo Scientific Chemicals

Catalog number: 138620250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 138620250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
95-15-8
IUPAC Name
1-benzothiophene
Molecular Formula
C8H6S
InChI Key
FCEHBMOGCRZNNI-UHFFFAOYSA-N
SMILES
S1C=CC2=CC=CC=C12
Appearance (Color)
White to yellow to pink to orange
Infrared spectrum
Conforms
Melting point
28°C to 33°C
Appearance (Form)
Crystalline solid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text