Thermo Scientific Chemicals

L(-)-Thiazolidine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 138640100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

L(-)-Thiazolidine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 138640100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 138640100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
34592-47-7
IUPAC Name
(4R)-1,3-thiazolidin-3-ium-4-carboxylate
Molecular Formula
C4H7NO2S
InChI Key
DZLNHFMRPBPULJ-VKHMYHEASA-N
SMILES
[O-]C(=O)[C@@H]1CSC[NH2+]1
Infrared spectrum
Conforms
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals or needles
Titration with NaOH
>=97.5 %
Specific optical rotation
-136° to -144° (20°C, 589 nm) (c=1, H2O)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text