Thermo Scientific Chemicals

Ácido L(-)-tiazolidina-4-carboxílico, 98 %, Thermo Scientific Chemicals

Número de catálogo: 138640250
25 g, Frasco de vidrio, Each
Thermo Scientific Chemicals

Ácido L(-)-tiazolidina-4-carboxílico, 98 %, Thermo Scientific Chemicals

Número de catálogo: 138640250
25 g, Frasco de vidrio, Each
Cantidad
Envase
Número de catálogo: 138640250
Precio (ARS)

Identificadores químicos

CAS
34592-47-7
IUPAC Name
(4R)-1,3-thiazolidin-3-ium-4-carboxylate
Molecular Formula
C4H7NO2S
InChI Key
DZLNHFMRPBPULJ-VKHMYHEASA-N
SMILES
[O-]C(=O)[C@@H]1CSC[NH2+]1
Infrared spectrum
Conforms
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals or needles
Titration with NaOH
>=97.5 %
Specific optical rotation
-136° to -144° (20°C, 589 nm) (c=1, H2O)

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text