p-Toluenethiol, 98%, Thermo Scientific Chemicals
p-Toluenethiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Toluenethiol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 138711000
Price (USD)/ Each
87.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
103.00 
Save 15.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
87.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
103.00 
Save 15.35 (15%)
Add to cart
p-Toluenethiol, 98%, Thermo Scientific Chemicals
Catalog number138711000
Price (USD)/ Each
87.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
103.00 
Save 15.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-methylbenzene-1-thiol
Molecular FormulaC7H8S
InChI KeyWLHCBQAPPJAULW-UHFFFAOYSA-N
SMILESCC1=CC=C(S)C=C1
Molecular Weight (g/mol)124.20
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Appearance (Form)Fused solid
GC>=97.5 %
Melting point40°C to 44°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only