Thermo Scientific Chemicals

p-Toluenethiol, 98%, Thermo Scientific Chemicals

Catalog number: 138715000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

p-Toluenethiol, 98%, Thermo Scientific Chemicals

Catalog number: 138715000
500 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
106-45-6
IUPAC Name
4-methylbenzene-1-thiol
Molecular Formula
C7H8S
InChI Key
WLHCBQAPPJAULW-UHFFFAOYSA-N
SMILES
CC1=CC=C(S)C=C1
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
Appearance (Form)
Fused solid
GC
>=97.5 %
Melting point
40°C to 44°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text